Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

2-Propanol 99.5+%

Kst.-Trommel, 5 L
Catalog # ACRO149320050
Irritant
Flammable
2-Propanol 99.5+%
Catalog # ACRO149320050
CAS Number:  67-63-0
2-Propanol 99.5+%
2-Propanol 99.5+%

Kst.-Trommel, 5 L

CAS Number:  67-63-0
Price
179.00 SFr.
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
  • CAS Number:67-63-0
  • Formula:(CH₃)₂CHOH
  • MDL Number:MFCD00011674
  • Molecular Weight:60.1 g/mol
  • Merck Index:13,05228
  • Boiling Pt:82 °C (1013 hPa)
  • Melting Pt:–89,5 °C
  • Density:0,785 g/cm³
  • Flash Pt:12 °C
  • Storage Temperature:Raumtemperatur
  • EINECS:200-661-7
  • UN:1219
  • ADR:3,II
  • REACH:01-2119457558-25

 

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.2 % (K.F.)
  • GC:
    ≥99.50 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.005 %

Specifications

  • Packung:
    Kst.-Trommel
  • VE:
    5 L

Specifications