(R)-(-)-Mandelsäure 99%
Glasflasche, 5 g
Anbieter: Thermo Fisher Scientific
(R)-(-)-Mandelsäure 99%
Katalognr. ACRO125320050
Anbieter: Thermo Fisher Scientific
CAS Nummer: 611-71-2
Direkt zu
Product Details & Documents
Synonyme: D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid
- CAS Nummer:611-71-2
- Formel:C₈H₈O₃
- MDL-Nummer:MFCD00064251
- Molekulargewicht:152.15 g/mol
- Siedepunkt:322 °C (1013 hPa)
- Schmelzender Platingehalt:130…135 °C
- Dichte:1,341 g/cm³ (20 °C)
- Blitz Pt:>190 °C
- Lagertemperatur:Raumtemperatur
- EINECS:210-276-6
Dokumente
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance:White to light yellow to light beige Crystalline powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:130 to 135 °C
- Water:≤1 % (K.F.)
- Sulfated ash:≤0.2 %
- Specific optical rotation:-152° to -158° (20 °C, 589 nm) (c=2, H2O)
- Titration with NaOH:≥98.5 %
Technische Daten
- Packung:Glasflasche
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



