5-Bromphthalid 98%
Glasflasche, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 5-Bromoisobenzofuran-1(3H)-one
- CAS Number:64169-34-2
- Formula:C₈H₅BrO₂
- MDL Number:MFCD01797360
- Molecular Weight:213.03 g/mol
- Melting Pt:162…166 °C
- Storage Temperature:Raumtemperatur
- EINECS:447-190-5
Specifications
Specification Test Results
- Appearance:Light yellow to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:162 to 166 °C
- HPLC:≥97.5 %
Specifications
- Packung:Glasflasche
- VE:25 g



