AuftragsverfolgungKontakt
Dip slides
Dip slides
Plate Count/Plate Count, 10
Katalognr. OXOIGFD07A
Dip slides
Katalognr. OXOIGFD07A
CAS Nummer:  
Dip slides
Plate Count/Plate Count, 10
CAS Nummer:  
Preis
33.70 SFr.
Einloggen für Verfügbarkeit
1 * 10 ST
Product Details & Documents
Simple and safe for an unskilled operator to use, these dip slides are coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective. This sterile dip slide can be inoculated in the ward or doctor’s surgery and then returned to its plastic container for easy transport back to the laboratory.
  • Make colony counting for the bacterial assessment simple
Documents
Technische Daten
Technische Daten
  • VE:
    10
  • Bezeichnung:
    Plate Count/Plate Count
Technische Daten
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.