Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Single tissue blots, multiple species
Human, mouse and rat spleen single tissue, multiple species blot, 1
Katalognr. TB36
Anbieter:   G-Biosciences
Single tissue blots, multiple species
Katalognr. TB36
Anbieter:  G-Biosciences
CAS Nummer:  
Single tissue blots, multiple species
Single tissue blots, multiple species
Human, mouse and rat spleen single tissue, multiple species blot, 1
CAS Nummer:  
Preis
SFr. 636.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Single tissue and multiple species blot contains four protein lanes with an identical GenLysate™ tissue lysate from human, mouse and rat species, and a protein marker lane.
  • Ready to screen
  • Single tissue multiple species (human, mouse, rat) blot
The premade blots are prepared with the GenLysate™ and then 50μg protein is loaded onto a 4 to 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane using Western Transfer Buffer. The blots are ready to be blocked and probed with the antibodies of choice.
Technische Daten
Technische Daten
  • VE:
    1
  • Bezeichnung:
    Human, mouse and rat spleen single tissue, multiple species blot
Technische Daten