Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Single tissue blots, multiple species
Single tissue blots, multiple species
Human, mouse and rat testis single tissue, multiple species blot, 1
Katalognr. TB32
Anbieter:   G-Biosciences
Single tissue blots, multiple species
Katalognr. TB32
Anbieter:  G-Biosciences
CAS Nummer:  
Single tissue blots, multiple species
Single tissue blots, multiple species
Human, mouse and rat testis single tissue, multiple species blot, 1
CAS Nummer:  
Preis
SFr. 636.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Single tissue and multiple species blot contains four protein lanes with an identical GenLysate™ tissue lysate from human, mouse and rat species, and a protein marker lane.
  • Ready to screen
  • Single tissue multiple species (human, mouse, rat) blot
The premade blots are prepared with the GenLysate™ and then 50μg protein is loaded onto a 4 to 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane using Western Transfer Buffer. The blots are ready to be blocked and probed with the antibodies of choice.
Technische Daten
Technische Daten
  • VE:
    1
  • Bezeichnung:
    Human, mouse and rat testis single tissue, multiple species blot
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.