Single tissue blots, multiple species
Human, mouse and rat lung single tissue, multiple species blot, 1
Katalognr. TB29
Anbieter: G-Biosciences
Direkt zu
Product Details & Documents
Single tissue and multiple species blot contains four protein lanes with an identical GenLysate™ tissue lysate from human, mouse and rat species, and a protein marker lane.
- Ready to screen
- Single tissue multiple species (human, mouse, rat) blot
The premade blots are prepared with the GenLysate™ and then 50μg protein is loaded onto a 4 to 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane using Western Transfer Buffer. The blots are ready to be blocked and probed with the antibodies of choice.
Technische Daten
Technische Daten
- VE:1
- Bezeichnung:Human, mouse and rat lung single tissue, multiple species blot