Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Proteomic protein controls
Proteomic protein controls
Protein proteomic control set, 4×2 mg, 1 SET
N° de catalogue PSC-05
Fournisseur:   G-Biosciences
Proteomic protein controls
N° de catalogue PSC-05
Fournisseur:  G-Biosciences
Numéro CAS:  
Proteomic protein controls
Protein proteomic control set, 4×2 mg, 1 SET
Numéro CAS:  
Prix
SFr. 397.00
Connexion Pour la disponibilité
1 * 1 SET
Product Details & Documents
Protein preparations specifically prepared for standardising electrophoresis methods and protocols. Substantially free from non-protein agents such as nucleic acids, detergents, salts, lipids and other common laboratory agents, the protein preparation has low conductivity (<50 μs). Supplied as dry protein pellets, simply re-hydrate and use. Proteomic protein sample controls are available for animal cells, E.coli, yeast, plant or as a control set that contains one 2 mg vial of each.
Spécifications
Spécifications
  • VE:
    1 SET
  • Bezeichnung:
    Protein proteomic control set, 4×2 mg
Spécifications
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.