Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Proteomic protein controls
Protein proteomic control, plant, 2×2 mg, 2
Catalog # PSC-04
Supplier:   G-Biosciences
Proteomic protein controls
Catalog # PSC-04
Supplier:  G-Biosciences
CAS Number:  
Proteomic protein controls
Protein proteomic control, plant, 2×2 mg, 2
CAS Number:  
Price
SFr. 189.00
Log in for availability
1 * 2 ST
Product Details & Documents
Protein preparations specifically prepared for standardising electrophoresis methods and protocols. Substantially free from non-protein agents such as nucleic acids, detergents, salts, lipids and other common laboratory agents, the protein preparation has low conductivity (<50 μs). Supplied as dry protein pellets, simply re-hydrate and use. Proteomic protein sample controls are available for animal cells, E.coli, yeast, plant or as a control set that contains one 2 mg vial of each.
Specifications
Specifications
  • VE:
    2
  • Bezeichnung:
    Protein proteomic control, plant, 2×2 mg
Specifications