AuftragsverfolgungKontakt
Proteomic protein controls | Avantor
Proteomic protein controls
Protein proteomic control, yeast, 2×2 mg, 2
Katalognr. PSC-01
Anbieter:   G-Biosciences
Proteomic protein controls
Katalognr. PSC-01
Anbieter:  G-Biosciences
CAS Nummer:  
Proteomic protein controls
Protein proteomic control, yeast, 2×2 mg, 2
CAS Nummer:  
Preis
SFr. 189.00
Einloggen für Verfügbarkeit
1 * 2 ST
Product Details & Documents
Protein preparations specifically prepared for standardising electrophoresis methods and protocols. Substantially free from non-protein agents such as nucleic acids, detergents, salts, lipids and other common laboratory agents, the protein preparation has low conductivity (<50 μs). Supplied as dry protein pellets, simply re-hydrate and use. Proteomic protein sample controls are available for animal cells, E.coli, yeast, plant or as a control set that contains one 2 mg vial of each.
Technische Daten
Technische Daten
  • VE:
    2
  • Bezeichnung:
    Protein proteomic control, yeast, 2×2 mg
Technische Daten