n-Hexan ≥95,0%, BAKER ANALYZED® HPLC, J.T.Baker®
Glasflasche, 5 L
Anbieter: Avantor
n-Hexan ≥95,0%, BAKER ANALYZED® HPLC, J.T.Baker®
Katalognr. 9304.5000
Anbieter: Avantor
CAS Nummer: 110-54-3
Direkt zu
Product Details & Documents
for use in High Performance Liquid Chromatography
Synonyme: Hexyl hydride, Hexanen, Hexane
- CAS Nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL-Nummer:MFCD00009520
- Molekulargewicht:86.18 g/mol
- UN:1208
- ADR:3,II
Documents
Zertifikate
Suche nach Chargennummerspec_BAKR9304.1000.pdf spec_BAKR9304.2500.pdf spec_BAKR9304.5000.pdf
Product specification
VWR J.T.Baker® High Purity solvents and reagents
Product Literature
Technische Daten
Ergebnisse des Spezifikationstests
- Assay (by GC):min. 95 %
- Residue after Evaporation:max. 2 ppm
- Substances Darkened by H2SO4:passes test
- Water (H2O):max. 0,01 %
- Ultraviolet Absorbance,(1.00-cm path vs water; curve smooth throughout stated range with no extraneous impurity peaks).:-
- at 210 nm:max. 0,2
- at 220 nm:max. 0,08
- at 254 nm:max. 0,008
- at 280 nm:max. 0,005
- at 350 nm:max. 0,005
- UV Cut-off, nm:max. 192
Technische Daten
- Packung:Glasflasche
- VE:5 L
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



