Dosen mit Deckel
180 mm, 180 mm
Katalognr. BOCH8624
Anbieter: BOCHEM
Direkt zu
Product Details & Documents
Jar made of stainless steel.
- Stackable
- Lid with handle
- Very good chemical and temperature resistance
Technische Daten
- Ext.Ø:180 mm
- Höhe:180 mm