Double bossheads
Double bosshead, heart screw M6, Edelstahl 18/10, 16,5 mm
BOCH5353
: BOCHEM
Double bossheads, either made of stainless steel or aluminium.
- Angle: 180 + 90°
- Different screw types available
- Bezeichnung:Double bosshead, heart screw M6
- Material:Edelstahl 18/10
- Für Stab Ø:16,5 mm