Cold traps with/without condensation drain
200 ml, with condensate drain
Katalognr. LENZ05355546
Anbieter: Lenz Laborglas
Direkt zu
Product Details & Documents
For direct filling with coolant. Double-walled design ensures low coolant consumption. Includes NS 29/32 standard ground socket for connecting to the apparatus and NS 29/32 standard ground cone for connecting to the vacuum pump.
DURAN®, klar.
- Sehr gute chemische Beständigkeit
- Hohe Temperaturbeständigkeit
- Minimale Wärmeausdehnung, dadurch bedingt hohe Temperaturwechselbeständigkeit
Dokumente
BSE TSE Glass Columns
BSE/TSE-Zertifikate
Technische Daten
- Bezeichnung:200 ml, with condensate drain
- Seq:04