BD® Blunt Needles
18 G, 40 mm, Stumpfe Fase, Sterile, Lila
Anbieter: BD Medical
Direkt zu
Product Details & Documents
BD blunt fill and blunt filter needles are specifically designed to reduce the risk of needlestick injuries during medication preparation.
- The blunt fill needle is designed for vial access and available with a BD Luer-Lok™ connection
- The blunt filter is designed for safe ampoule medication preparation
- BD blunt fill needles have female luer fittings, which mate to male luer fittings
- BD blunt filter needle has a 5-micron filter for particles such as glass and plastic fragments, for safer medication delivery
- Natural rubber latex is not part of the material formulation
- Sterile, singe use
Achtung! Bitte beachten: Note: The BD blunt fill and filter needles are not for skin injection.
Technische Daten
- Farbcode:Lila
- Gauge:18 G
- Typ:Sterile
- Spitzentyp:Stumpfe Fase
- Länge:40 mm
- Seq:01
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.