AuftragsverfolgungKontakt
Quarz-Kontrollplatten
Standardkontrollplatte aus Quarz, −30 °Z, −10 °A, 1
Katalognr. 635-0405
Anbieter:   BELLINGHAM STANLEY
Quarz-Kontrollplatten
Katalognr. 635-0405
Anbieter:  BELLINGHAM STANLEY
CAS Nummer:  
Quarz-Kontrollplatten
Quarz-Kontrollplatten
Standardkontrollplatte aus Quarz, −30 °Z, −10 °A, 1
CAS Nummer:  
Preis
SFr. 1’790.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Quartz control plates for the calibration and verification of polarimeters. of optical and digital polarimeters and saccharimeters.
  • Inert quartz with relatively low temperature coefficient, easy to handle, and infinite shelf life
  • Available in three optical rotation values
  • Three decimal place uncertainty
The optional temperature saddles may used as an interface between the quartz plate and instrument temperature sensor to provide fine compensation.
 : Traceable to PTB. UKAS-MRA calibration in accordance with ISO 17025:2005 between 325-880nm available.
 : Sadlles can be ordered in option.
 : Supplied with a certificate of verification in a plastic storage case.
Technische Daten
Technische Daten
  • VE:
    1
  • Bezeichnung:
    Standardkontrollplatte aus Quarz, −30 °Z, −10 °A