Sulfolan ≥99%
Glass bottle for solids, 5.000 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 126-33-0 Molecular Formula: C4H8O2S Formula Weight: 120.17 Flash Point: 166°(330°F) Refractive Index: 1.4825 Odour: Weak, characteristic Melting Point: 25-28° Boiling Point: 285° Beilstein Reference: 107765 Merck Reference: 14,8955 MDL No.: MFCD00005484
Synonyme: Tetrahydrothiophene-1,1-dioxide, Tetramethylene sulphone, Tetrahydrothiophene 1,1-dioxide
- CAS Nummer:126-33-0
- Formel:C₄H₈O₂S
- MDL-Nummer:MFCD00005484
- Molekulargewicht:120.17 g/mol
- Merck-Index:14,8955
- Siedepunkt:285 °C
- Schmelzender Platingehalt:25…28 °C
- Dichte:1,261
- Blitz Pt:166 °C (330 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:204-783-1
Documents
Zertifikate
Suche nach ChargennummerOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Stockroom Essentials
Product Literature
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:5.000 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



