Extraction thimbles
58×170 mm
Katalognr. 537-0012
Anbieter: PRAT DUMAS
Direkt zu
Product Details & Documents
Pure cotton cellulose
- Neutral and free of fats and grease
- Moulded in one piece
- Very resistant to solvents
Applications include extraction of sludges, asphalt, foodstuffs and in the analysis and quality control of powders and dusts.
Technische Daten
- I-Ø×H:58×170 mm