Test kits, DEHA, CHECKIT®
DEHA test kit, 0 - 0,5 mg/L DEHA, 1 KIT
Katalognr. LOVI147370
Anbieter: Lovibond
Direkt zu
Product Details & Documents
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
- Easy operation and exact reagent dosing
- Tablet reagents have minimum guaranteed shelf life of 5/10 years
The operating instructions provide a step-by-step explanation of conducting the water test to achieve reliable and accurate measurements in the minimum of time.
Lieferinformationen: Test kit includes CHECKIT® discs, cells, stirring rod and AQUALYTIC® tablet reagents (for 30 tests) for the desired test supplied in a sturdy plastic case.
Technische Daten
Technische Daten
- VE:1 KIT
- Bezeichnung:DEHA test kit
- Messbereich:0 - 0,5 mg/L DEHA
Technische Daten
Accessories
Produktinformationen
- Katalognr.BezeichnungVEVerfügbarkeitPreis
- LOVI145000CHECKIT® Comparator, path length 13,5 mm1Einloggen für Verfügbarkeit1 * 1 ST - SFr. 27.70
Produktinformationen
- Katalognr.BezeichnungVEVerfügbarkeitPreis
- LOVI145500CHECKIT® plastic cells, pack of 10, with lid, 10 ml, 13,5 mm path length10Einloggen für Verfügbarkeit1 * 10 ST - SFr. 62.80
Produktinformationen
- Katalognr.BezeichnungVEVerfügbarkeitPreis
- LOVI145505CHECKIT® plastic cells, pack of 5, with lid, 10 ml, 13,5 mm path length5Einloggen für Verfügbarkeit1 * 5 ST - SFr. 37.10
Produktinformationen
- Katalognr.BezeichnungVEVerfügbarkeitPreis
- LOVI145510CHECKIT® plastic cells, pack of 100, with lid, 10 ml, 13,5 mm path length100Einloggen für Verfügbarkeit1 * 100 ST - SFr. 520.00