Watch glasses
Watch glass, glass, with ground edges, 100 mm
Katalognr. MURRG850/03
Anbieter: MURRAY S
Direkt zu
Product Details & Documents
Made of glass.
- Good chemical and physical properties
- Use in applications that do not involve high thermal stresses
- Quick and easy to clean
With ground edges for covering beakers.
Technische Daten
- Ø:100 mm
- Bezeichnung:Watch glass, glass, with ground edges
- Seq:03