Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Watch glasses | Avantor
Watch glasses
Watch glass, glass, with ground edges, 75 mm
Katalognr. MURRG850/02
Anbieter:   MURRAY S
Watch glasses
Katalognr. MURRG850/02
Anbieter:  MURRAY S
Watch glasses
Watch glass, glass, with ground edges, 75 mm
Preis
SFr. 74.40
Einloggen für Verfügbarkeit
1 * 10 ST
Product Details & Documents
Made of glass.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean
With ground edges for covering beakers.
Technische Daten
  • Ø:
    75 mm
  • Bezeichnung:
    Watch glass, glass, with ground edges
  • Seq:
    02