Magnetrührscheiben
, 9,92 mm, 19,76 mm
KART778
: KARTELL
PTFE-beschichtete Magnetrührstäbe mit konvexer Form für optimale Zentrierung.
- Effektives Rühren
- Für Lebensmittel geeignet
Certifications:
Delivery information:
Packaging:
Caution:
- Ø:19,76 mm
- Länge:9,92 mm
- Seq:02
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.