Filtertiegel
50 ml, 3, 40 mm
LENZ07185340
Filtration crucibles made of high quality borosilicate glass 3,3.
- Platte Ø:40 mm
- Porosität:3
- Kapazität:50 ml
- Seq:11
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



