Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Overhead stirrer tools, anchor stirrers | Avantor

Overhead stirrer tools, anchor stirrers

Anchor stirrer, DURAN®, 100 LF, 10000 ml, 16 mm
Katalognr. LENZ06321088
Anbieter:   Lenz Laborglas
Overhead stirrer tools, anchor stirrers
Katalognr. LENZ06321088
Anbieter:  Lenz Laborglas
Overhead stirrer tools, anchor stirrers
Overhead stirrer tools, anchor stirrers

Anchor stirrer, DURAN®, 100 LF, 10000 ml, 16 mm

Preis
SFr. 195.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Anchor stirrers with GL thread connection.

  • Made of precision calibrated glass tube
  • With ground and polished bearing surface
  • Suitable for flange vessels

Dokumente

Technische Daten
  • Bezeichnung:
    Anchor stirrer, DURAN®, 100 LF
  • Schaft - Ø:
    16 mm
  • Für Gefäße:
    10000 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.