Kühler, Davies
400 mm, 24/29 NS, 24/29 NS
Catalog # LENZ05215413
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN® borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With double sized coiling area. Water connection with two GL 14 threaded plastic olives.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Kern:24/29 NS
- Mantellänge:400 mm
- Hülse:24/29 NS
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.