AuftragsverfolgungKontakt
Kühler, Davies
400 mm, 24/29 NS, 24/29 NS
Catalog # LENZ05215413
Supplier:   Lenz Laborglas
Kühler, Davies
Catalog # LENZ05215413
Supplier:  Lenz Laborglas
Kühler, Davies
Kühler, Davies
400 mm, 24/29 NS, 24/29 NS
Price
251.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN® borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With double sized coiling area. Water connection with two GL 14 threaded plastic olives.
Documents
Specifications
  • Kern:
    24/29 NS
  • Mantellänge:
    400 mm
  • Hülse:
    24/29 NS