Spore strips, Attest™
Bacillus-subtilis-Sporenstreifen, Ethylenoxid-Sterilisation
Katalognr. MMMEM1264
Anbieter: 3M MEDICAL
Direkt zu
Product Details & Documents
3M™ Attest™ Biological Monitoring Systems are true biological indicators, in self-contained indicator capsules. They enable checks on the biological reliability and performance of each steriliser without laboratory facilities and laboratory trained personnel.
- Self-contained, reducing the risk of false positives due to contamination
- Colour change gives clear unambiguous, visual indication of PASS or FAIL
- Time saving and results in 24 to 48 hours which require no interpretation
Dokumente
Zertifikate
Suche nach ChargennummerCoC_MMMEM1262_3394W6 CoC_MMMEM1262_33FTPR CoC_MMMEM1262_33JKDH CoC_MMMEM1262_33LMPF MMMEM1262_343F4T.pdf
MMMEM1262_TDS_EN
Technisches Datenblatt
Technische Daten
- Bezeichnung:Bacillus-subtilis-Sporenstreifen, Ethylenoxid-Sterilisation