Magnetic stirring bars, plain, PTFE
50 mm, 7 mm
Anbieter: COWIE
Direkt zu
Product Details & Documents
Plain PTFE magnetic stirring bars have similar performance to cylindrical bars, but provide more turbulence at low speeds.
Technische Daten
- Ø:7 mm
- Länge:50 mm
- Seq:07



