Septum caps, Nalgene®
24 - 415
Catalog # NALG342178-0240
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
HDPE with silicone/PTFE septum.
- Sterile 10⁻⁶ SAL
- No adhesives used to attach septum to high density polyethylene closure
- Non-cytotoxic and non-pyrogenic
- Dual septum material for improved gas barrier
Specifications
- Verschlussgröße:24 - 415
- Cat. No.:NALG342178-0240
- Umweltzertifizierung:NAR Certificate of Retirement
- Umweltfreundlichkeit:Umweltschonende Herstellung