Abdeckmatten für Deep-Well-Platten
Verschlussmatten, PP, für 96-Well-Platten (0,5 ml)
391-5713
: Brand
Cover mats reduce the maximum volume of wells.
- Bezeichnung:Verschlussmatten, PP, für 96-Well-Platten (0,5 ml)
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.