Kresolrot
Glasflasche für Feststoffe, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 1733-12-6 Molecular Formula: C21H18O5S Formula Weight: 382.44 Melting Point: 300° Beilstein Reference: 343399 Merck Reference: 14,2578 MDL No.: MFCD00005878
Synonyms: o-Cresolsulphonphthalein, 3,3-Bis(4-hydroxy-3-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:1733-12-6
- Formula:C₂₁H₁₈O₅S
- MDL Number:MFCD00005878
- Molecular Weight:382.43 g/mol
- Merck Index:14,2578
- Melting Pt:>300 °C
- Storage Temperature:Raumtemperatur
- EINECS:217-064-2
Documents
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:100 g



