Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
β-Alanin ≥98%

β-Alanin ≥98%

Glasflasche für Feststoffe, 1.000 g
Catalog # A16665.0B
β-Alanin ≥98%
Catalog # A16665.0B
CAS Number:  107-95-9
β-Alanin ≥98%
β-Alanin ≥98%

Glasflasche für Feststoffe, 1.000 g

CAS Number:  107-95-9
Price
184.00 SFr.
Log in for availability
1 * 1.000 g
Product Details & Documents

Purity: 98% CAS No.: 107-95-9 Molecular Formula: C3H7NO2 Formula Weight: 89.09 Physical Form: Cryst powder Density: 1.437 Appearance: White Odour: Odourless Melting Point: ca 205° dec. Beilstein Reference: 906793 Merck Reference: 14,205 MDL No.: MFCD00008200

Synonyms:  H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
  • CAS Number:107-95-9
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00008200
  • Molecular Weight:89.09 g/mol
  • Merck Index:14,205
  • Boiling Pt:201 °C (1013 hPa)
  • Melting Pt:∼ 205 °C (Zers.)
  • Melting Pt:∼ 205 °C (Zersetzung)
  • Density:1,437
  • Storage Temperature:Raumtemperatur
  • EINECS:203-536-5
Specifications

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    1.000 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.