2-Mercaptoethanol ≥98%
Glasflasche für Feststoffe, 1.000 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 98+% CAS No.: 60-24-2 Molecular Formula: HSCH2CH2OH Formula Weight: 78.13 Flash Point: 73°(163°F) Refractive Index: 1.500 Odour: Unpleasant Melting Point: -100° Boiling Point: 156-158° Beilstein Reference: 773648 Merck Reference: 14,5869 UN No.: 2966 MDL No.: MFCD00004890
Synonyme: 2-Hydroxyethyl mercaptan, β-Mercaptoethanol
- CAS Nummer:60-24-2
- Formel:HSCH₂CH₂OH
- MDL-Nummer:MFCD00004890
- Molekulargewicht:78.14 g/mol
- Merck-Index:14,5869
- Siedepunkt:156…158 °C
- Schmelzender Platingehalt:–100 °C
- Dichte:1,114
- Blitz Pt:73 °C (163 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:200-464-6
- UN:2966
- ADR:6.1,II
Dokumente
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:1.000 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



