(±)-Epichlorhydrin ≥99%
Glasflasche für Feststoffe, 2.500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 106-89-8 Molecular Formula: C3H5ClO Formula Weight: 92.53 Flash Point: 32°(90°F) Physical Form: Liquid Density: 1.182 Refractive Index: 1.4380 Appearance: Colourless Odour: Like chlorine Melting Point: -57° Boiling Point: 115-117° Beilstein Reference: 79785 Merck Reference: 14,3611 UN No.: 2023 MDL No.: MFCD00005132
A cyclodextrin cross-linking reagent
Synonyms: (±)-1-Chloro-2,3-epoxypropane, Epichlorohydrin, (±)-3-Chloro-1,2-epoxypropane, 1-Chloro-2,3-epoxypropane, Epichlorhydrin, (±)-Glycerol epichlorhydrin, 3-Chloro-1,2-epoxypropane
- CAS Number:106-89-8
- Formula:C₃H₅ClO
- MDL Number:MFCD00005132
- Molecular Weight:92.52 g/mol
- Merck Index:14,3611
- Boiling Pt:115…117 °C
- Melting Pt:–57 °C
- Density:1,182
- Flash Pt:32 °C (90 °F)
- Storage Temperature:Raumtemperatur
- EINECS:203-439-8
- UN:2023
- ADR:6.1,II
Documents
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:2.500 g



