Methyldecanoat ≥99%
Glasflasche für Feststoffe, 100 g
Purity: 99% CAS No.: 110-42-9 Molecular Formula: CH3(CH2)8CO2CH3 Formula Weight: 186.30 Flash Point: 94°(201°F) Refractive Index: 1.4250 Melting Point: -18° Boiling Point: 223-225° Beilstein Reference: 1759170 MDL No.: MFCD00009580
: Decanoic acid methyl ester, Capric acid methyl ester, Decylic acid methyl ester, Methyl caprate
- :110-42-9
- :C₁₁H₂₂O₂
- :MFCD00009580
- :186.29 g/mol
- :223…225 °C
- :–18 °C
- :0,877
- :94 °C (201 °F)
- :Raumtemperatur
- :203-766-6
- Packung:Glasflasche für Feststoffe
- VE:100 g