Phthalid ≥98%
Glass bottle for solids, 500 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 98+% CAS No.: 87-41-2 Molecular Formula: C8H6O2 Formula Weight: 134.13 Flash Point: 152°(305°F) Melting Point: 72-75° Boiling Point: 289-291° Beilstein Reference: 114632 MDL No.: MFCD00005906
Synonyme: Isobenzofuran-1(3H)-one
- CAS Nummer:87-41-2
- Formel:C₈H₆O₂
- MDL-Nummer:MFCD00005906
- Molekulargewicht:134.13 g/mol
- Siedepunkt:289…291 °C
- Schmelzender Platingehalt:72…75 °C
- Dichte:1,1
- Blitz Pt:152 °C (305 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:201-744-0
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:500 g



