Picolinamid ≥98%
Glass bottle for solids, 25 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 98% CAS No.: 1452-77-3 Molecular Formula: C6H6N2O Formula Weight: 122.13 Melting Point: 104-108° Boiling Point: 143°/20mm MDL No.: MFCD00023483
Synonyme: 2-Pyridinecarboxamide, Pyridine-2-carboxamide
- CAS Nummer:1452-77-3
- Formel:C₆H₆N₂O
- MDL-Nummer:MFCD00023483
- Molekulargewicht:122.13 g/mol
- Siedepunkt:143 °C (20 mmHg)
- Schmelzender Platingehalt:104…108 °C
- Lagertemperatur:Raumtemperatur
- EINECS:215-921-5
Documents
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:25 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



