Hydroxylammoniumchlorid 99%
Glasflasche für Feststoffe, 500 g
Supplier: Thermo Fisher Scientific
Hydroxylammoniumchlorid 99%
Catalog # A15398.36
Supplier: Thermo Fisher Scientific
CAS Number: 5470-11-1
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 5470-11-1 Molecular Formula: H3NO.HCl Formula Weight: 69.49 Odour: Acidic pH: 3.6 Melting Point: 152° dec. Beilstein Reference: 3539763 Merck Reference: 14,4828 UN No.: 2923 MDL No.: MFCD00051089
Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride
- CAS Number:5470-11-1
- Formula:NH₂OH·HCl
- MDL Number:MFCD00051089
- Molecular Weight:69.49 g/mol
- Merck Index:14,4828
- Boiling Pt:306 °C (1013 hPa)
- Melting Pt:152 °C (Zers.)
- Melting Pt:152 °C (Zersetzung)
- Density:1,67
- Storage Temperature:Raumtemperatur
- EINECS:226-798-2
- UN:2923
- ADR:8,III
Documents
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



