(4-Hydroxyphenyl)essigsäure ≥99%
Glass bottle for solids, 25 g
Purity: 99% CAS No.: 156-38-7 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 148-150° Beilstein Reference: 1448766
: 4-Hydroxyphenylacetic Acid
- :156-38-7
- :(C₆H₄(OH))CH₂COOH
- :MFCD00004347
- :152.15 g/mol
- :347 °C (1013 hPa)
- :148…150 °C
- :Raumtemperatur
- :205-851-3
- Packung:Glass bottle for solids
- VE:25 g
- Cat. No.:A15018.14