Bicin ≥99%
Glasflasche für Feststoffe, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 150-25-4 Molecular Formula: C6H13NO4 Formula Weight: 163.17 pH: 4-5 Melting Point: ca 191° dec. Beilstein Reference: 1769362 Merck Reference: 14,1201 MDL No.: MFCD00004295
Good's buffers
Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine
- CAS Number:150-25-4
- Formula:C₆H₁₃NO₄
- MDL Number:MFCD00004295
- Molecular Weight:163.17 g/mol
- Merck Index:14,1201
- Melting Pt:∼ 191 °C (Zers.)
- Melting Pt:∼ 191 °C (Zersetzung)
- Storage Temperature:Raumtemperatur
- EINECS:205-755-1
Documents
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



