2-Methylbenzophenon ≥98%
50 g
Purity: 98+% CAS No.: 131-58-8 Molecular Formula: C14H12O Formula Weight: 196.25 Flash Point: 110°(230°F) Refractive Index: 1.5950 Melting Point: -18° Boiling Point: 309-311° Beilstein Reference: 2045469 Merck Reference: 14,7317 MDL No.: MFCD00008518
: o-Methylbenzophenone
- :131-58-8
- :C₁₄H₁₂O
- :MFCD00008518
- :196.25 g/mol
- :14,7317
- :309…311 °C
- :–18 °C
- :1,085
- :>110 °C (230 °F)
- :Raumtemperatur
- :205-032-0
- VE:50 g