Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
2-Methylbenzophenon ≥98%
50 g
  A14779.18
2-Methylbenzophenon ≥98%
  A14779.18
 :  131-58-8
2-Methylbenzophenon ≥98%
2-Methylbenzophenon ≥98%
50 g
 :  131-58-8
254.00 SFr.
1 * 50 g
Purity: 98+% CAS No.: 131-58-8 Molecular Formula: C14H12O Formula Weight: 196.25 Flash Point: 110°(230°F) Refractive Index: 1.5950 Melting Point: -18° Boiling Point: 309-311° Beilstein Reference: 2045469 Merck Reference: 14,7317 MDL No.: MFCD00008518
 :  o-Methylbenzophenone
  •  :131-58-8
  •  :C₁₄H₁₂O
  •  :MFCD00008518
  •  :196.25 g/mol
  •  :14,7317
  •  :309…311 °C
  •  :–18 °C
  •  :1,085
  •  :>110 °C (230 °F)
  •  :Raumtemperatur
  •  :205-032-0
  • VE:
    50 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.