Inosin ≥98%
Glass bottle for solids, 5 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Applications: Suppresses the increase of glucose and insulin used in blood.
Synonyme: 9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one
- CAS Nummer:58-63-9
- Formel:C₁₀H₁₂N₄O₅
- MDL-Nummer:MFCD00066770
- Molekulargewicht:268.23 g/mol
- Merck-Index:14,4975
- Schmelzender Platingehalt:212…215 °C (Zers.)
- Schmelzender Platingehalt:212…215 °C (Zersetzung)
- Lagertemperatur:Raumtemperatur
- EINECS:200-390-4
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



