L(-)-Cystin ≥99%
Glass bottle for solids, 50 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 56-89-3 Molecular Formula: C6H12N2O4S2 Formula Weight: 240.30 Physical Form: Cryst powder Density: 1.68 Appearance: White Melting Point: 240° dec. Beilstein Reference: 1728094 Merck Reference: 14,2782 Optical Rotation: -219° (c=1 in 1N HCl) MDL No.: MFCD00064228
Synonyme: (R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2
- CAS Nummer:56-89-3
- Formel:HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
- MDL-Nummer:MFCD00064228
- Molekulargewicht:240.3 g/mol
- Merck-Index:14,2782
- Siedepunkt:428 °C (1013 hPa)
- Schmelzender Platingehalt:>240 °C (Zers.)
- Schmelzender Platingehalt:>240 °C (Zersetzung)
- Dichte:1,68
- Lagertemperatur:Raumtemperatur
- EINECS:200-296-3
Dokumente
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:50 g



