5-Bromisatin ≥90%
Glass bottle for solids, 50 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 90+% CAS No.: 87-48-9 Molecular Formula: C8H4BrNO2 Formula Weight: 226.03 Melting Point: ca 250° dec. Beilstein Reference: 383760
Synonyme: 5-Bromoindoline-2,3-dione, 5-Bromo-1H-indole-2,3-dione
- CAS Nummer:87-48-9
- Formel:C₈H₄BrNO₂
- MDL-Nummer:MFCD00149345
- Molekulargewicht:226.03 g/mol
- Schmelzender Platingehalt:∼ 250 °C (Zers.)
- Schmelzender Platingehalt:∼ 250 °C (Zersetzung)
- Lagertemperatur:Raumtemperatur
- EINECS:201-747-7
Documents
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:50 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



