Phthalaldehyd ≥98%
Glasflasche für Feststoffe, 10 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 98% CAS No.: 643-79-8 Molecular Formula: C8H6O2 Formula Weight: 134.14 Flash Point: 132°(269°F) Melting Point: 53-57° Beilstein Reference: 878317 UN No.: 2923 MDL No.: MFCD00003335
Synonyme: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde
- CAS Nummer:643-79-8
- Formel:C₈H₆O₂
- MDL-Nummer:MFCD00003335
- Molekulargewicht:134.13 g/mol
- Siedepunkt:266 °C (1013 hPa)
- Schmelzender Platingehalt:53…57 °C
- Dichte:1,13 g/cm³ (20 °C)
- Blitz Pt:132 °C (269 °F)
- Lagertemperatur:Kühlschrank
- EINECS:211-402-2
- UN:2923
- ADR:8,II
Documents
Zertifikate
Suche nach ChargennummerALFAA13299.09_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:10 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



