γ-(N-Morpholino)propansulfonsäure (MOPS)
Glass bottle for solids, 250 g
Katalognr. GRBU1081.0250
Marketsource
Anbieter: GERBU BIOCHEMICALS
γ-(N-Morpholino)propansulfonsäure (MOPS)
Katalognr. GRBU1081.0250
Anbieter: GERBU BIOCHEMICALS
Marketsource
CAS Nummer: 1132-61-2
Direkt zu
Product Details & Documents
Synonyme: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
- CAS Nummer:1132-61-2
- Formel:C₇H₁₅NO₄S
- MDL-Nummer:MFCD00006183
- Molekulargewicht:209.27 g/mol
- Merck-Index:13,06288
- Schmelzender Platingehalt:> 250 °C
- Blitz Pt:110 °C (geschlossener Tiegel)
- Lagertemperatur:Raumtemperatur
- EINECS:214-478-5
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:250 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



