γ-(N-Morpholino)propansulfonsäure (MOPS)
Glass bottle for solids, 100 g
GRBU1081.0100
Marketsource
: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
- :1132-61-2
- :C₇H₁₅NO₄S
- :MFCD00006183
- :209.27 g/mol
- :13,06288
- :> 250 °C
- :110 °C (geschlossener Tiegel)
- :Raumtemperatur
- :214-478-5
- Packung:Glass bottle for solids
- VE:100 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.