Dithiooxamid ≥98%
Glasflasche für Feststoffe, 25 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 98% CAS No.: 79-40-3 Molecular Formula: C2H4N2S2 Formula Weight: 120.2 Melting Point: 245° dec. MDL No.: MFCD00004941
Synonyme: Ethanebis(thioamide), Rubeanic acid
- CAS Nummer:79-40-3
- Formel:C₂H₄N₂S₂
- MDL-Nummer:MFCD00004941
- Molekulargewicht:120.2 g/mol
- Schmelzender Platingehalt:245 °C (Zers.)
- Schmelzender Platingehalt:245 °C (Zersetzung)
- Lagertemperatur:Raumtemperatur
- EINECS:201-203-9
Documents
Technische Daten
Technische Daten
- Packung:Glasflasche für Feststoffe
- VE:25 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



