Chlorphenolrot
Glass bottle for solids, 25 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
CAS No.: 4430-20-0 Molecular Formula: C19H12Cl2O5S Formula Weight: 432.28 Beilstein Reference: 354053 MDL No.: MFCD00005877
Synonyme: 3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
- CAS Nummer:4430-20-0
- Formel:C₁₉H₁₂Cl₂O₅S
- MDL-Nummer:MFCD00005877
- Molekulargewicht:423.27 g/mol
- Schmelzender Platingehalt:∼261 °C
- Lagertemperatur:Raumtemperatur
- EINECS:224-619-2
Dokumente
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:25 g