Methyllaurat ≥99%
25 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Purity: 99% CAS No.: 111-82-0 Molecular Formula: CH3(CH2)10CO2CH3 Formula Weight: 214.35 Flash Point: 125°(257°F) Refractive Index: 1.4325 Melting Point: 4-5° Boiling Point: 261-262° Beilstein Reference: 1767780 MDL No.: MFCD00008966
Synonyme: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester
- CAS Nummer:111-82-0
- Formel:C₁₃H₂₆O₂
- MDL-Nummer:MFCD00008966
- Molekulargewicht:214.35 g/mol
- Siedepunkt:261…262 °C
- Schmelzender Platingehalt:4…5 °C
- Dichte:0,870
- Blitz Pt:125 °C (257 °F)
- Lagertemperatur:Raumtemperatur
- EINECS:203-911-3
Dokumente
Zertifikate
Suche nach Chargennummermsds_ALFAA12492
Sicherheitsdatenblatt (SDS)
Organic Building Blocks and Intermediates
Anwendungshinweise
Technische Daten
Technische Daten
- VE:25 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



