Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Methyllaurat ≥99%

Methyllaurat ≥99%

Glasflasche für Feststoffe, 5 g
Catalog # A12492.06
Methyllaurat ≥99%
Catalog # A12492.06
CAS Number:  111-82-0
Methyllaurat ≥99%
Methyllaurat ≥99%

Glasflasche für Feststoffe, 5 g

CAS Number:  111-82-0
Price
80.80 SFr.
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 99% CAS No.: 111-82-0 Molecular Formula: CH3(CH2)10CO2CH3 Formula Weight: 214.35 Flash Point: 125°(257°F) Refractive Index: 1.4325 Melting Point: 4-5° Boiling Point: 261-262° Beilstein Reference: 1767780 MDL No.: MFCD00008966

Synonyms:  Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester
  • CAS Number:111-82-0
  • Formula:C₁₃H₂₆O₂
  • MDL Number:MFCD00008966
  • Molecular Weight:214.35 g/mol
  • Boiling Pt:261…262 °C
  • Melting Pt:4…5 °C
  • Density:0,870
  • Flash Pt:125 °C (257 °F)
  • Storage Temperature:Raumtemperatur
  • EINECS:203-911-3
Specifications

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.